BioClean Alpha™ Sterile Latex Gloves
BioClean Alpha™ Sterile Latex Gloves
Actual product may vary
BioClean Alpha™ Sterile Latex Gloves
Provides exceptional flexibility and is anatomically shaped for enhanced wearer comfort. Nitritex™ BioClean Alpha™ Sterile Latex Gloves features an extra-thick beaded cuff adds strength and stability on the arm, and reduces the risk of the gloves tearing when donning.
Have Questions?
Catalog NumberSize
NRTAL300-707
Catalog number NRTAL300-70
Price (SGD)
527.00
CS
Add to cart
Size:
7
Price (SGD)
527.00
CS
Add to cart
  • Extra thick beaded cuff to reduce tearing when donning
  • Powder-free EasyOn™ proprietary technology allows for easy double-donning
  • Non-particulating EasyTear™ packaging
  • Mid-arm 300mm (12 in.) length provides extra protection
  • Latex gloves offer exceptional flexibility and comfort
  • Anatomically-shaped, hand-specific gloves for a close fit
Specifications
Certifications/Compliance (Europe)ISO 9001:2015, (EU)2016/425, EN420:2003 +A1:2009 and EN374-1:2016, EN374-5:2016 and EN421:2010, EN455-2:2015, ISO 11137-1:2006, (EC)1935/2004 and (EC) 2023/2006
Cleanroom ClassClass 10 (ISO 4)
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
GradeEU GMP Grade A
HandSpecific
MaterialLatex
PowderedPowder Free
Product TypeCleanroom Gloves
Size7
SterilitySterile
Glove Cuff StyleBeaded/Rolled
Packaging TypeBagged
QuantityOne pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs)
Unit SizeCS