Nunc™ Lab-Tek™ II Chambered Coverglass
Actual product may vary
Thermo Scientific
Nunc™ Lab-Tek™ II Chambered Coverglass
Culture and view living cells on a single device. This coverglass has optimum optical characteristics for high-magnification microscopy and confocal image analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsMaterialDescription
NNU1554098Borosilicate Glass (1.5)8-well Chambered Coverglass w/ non-removable wells
FIS12-565-3351Borosilicate Glass (1.5)1-well Chambered Coverglass w/ non-removable well
NNU1553792Borosilicate Glass (1.5)2-well Chambered Coverglass w/ non-removable wells
FIS12-565-3374Borosilicate Glass (1.5)4-well Chambered Coverglass w/ non-removable wells
NNU1553824Borosilicate Glass (1.5)4-well Chambered Coverglass w/ non-removable wells
FIS12-565-3388Borosilicate Glass (1.5)8-well Chambered Coverglass w/ non-removable wells
Catalog number NNU155409
Price (SGD)
-
Wells:
8
Material:
Borosilicate Glass (1.5)
Description:
8-well Chambered Coverglass w/ non-removable wells
Culture, examine, and analyze living cells on a single device. Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass with a No. 1.5 borosilicate glass bottom offers the ideal optical characteristics required for high-magnification microscopy and confocal image analysis. Available in 1-, 2-, 4-, and 8-well formats. The upper chamber structure cannot be removed from the coverglass.
Features:
  • Media chamber securely mounted to the No. 1.5 borosilicate coverglass by biocompatible acrylic adhesive
  • The polystyrene chamber is not removable and requires inverted microscope for viewing
  • Gas exchange achieved with minimal evaporation through tabbed polystyrene cover
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Wells8
Culture Area0.7 cm2
Lid MaterialPolystyrene
Description8-well Chambered Coverglass w/ non-removable wells
Surface TreatmentCell Culture
MaterialBorosilicate Glass (1.5)
Volume (Metric) Working0.2 to 0.5 mL
Thickness (Metric)0.16 to 0.19 mm
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack16
Dimensions (LxW)55 x 24 mm
SterilitySterile
Closure ColorClear
Unit SizePack