Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Actual product may vary
Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Grow cells directly on slides with the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWellsMaterialDescription
NNU1545348RS Glass8-well Chamber Slide w/ removable wells
NNU1544612RS Glass2-well Chamber Slide w/ removable wells
NNU1545264RS Glass4-well Chamber Slide w/ removable wells
FKS102341218RS Glass8-well Chamber Slide w/ removable wells
Catalog number NNU154534
Price (SGD)
-
Wells:
8
Material:
RS Glass
Description:
8-well Chamber Slide w/ removable wells
Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features:
  • Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
  • Tabbed polystyrene lid allows gas exchange with minimal evaporation.
  • Removal tool separates media chamber from glass slide.
  • Biocompatible adhesive hydrophobic well border is printed on slide.
  • Slide features a white frosted writing area.
Includes:
  • Chamber removal tool
Ordering Information:
  • Can be purchased in case of 96 or pack of 16.
  • Shipped sterile in convenient stackable incubation trays.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Wells8
Culture Area0.7 cm2
Lid MaterialPolystyrene
Description8-well Chamber Slide w/ removable wells
Surface TreatmentCell Culture
MaterialRS Glass
Volume (Metric) Working0.2 to 0.5 mL
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack16
Dimensions (LxW)74 x 25 mm
SterilitySterile
Closure ColorClear
Unit SizePack