Search Thermo Fisher Scientific
Catalog Number | Bottom Shape | Capacity (Metric) | Thread Style | Style | Unit Size |
---|---|---|---|---|---|
NNC363401 | Round | 1.8 mL | Internal | Writing Surface | Pack |
374080 | Conical (Self-standing) | 0.5 mL | Internal | No Writing Surface, A-R Base | CS |
375353 also known as NNC375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base | PK |
FIS12-565-164N | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base | PK |
FKS10544641 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base | PK |
NNC375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base | Pack |
FIS12-565-297 | Conical (Self-standing) | 1 mL | External | No Writing Surface, A-R Base | PK |
377224 also known as NNC377224 | Conical (Self-standing) | 1 mL | Internal | Writing Surface, A-R Base | PK |
NNC377224 | Conical (Self-standing) | 1 mL | Internal | Writing Surface, A-R Base | Pack |
366656 also known as NNC366656 | Conical (Self-standing) | 1 mL | Internal | Writing Surface | PK |
375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base | CS |
NNC375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base | Pack |
347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base | PK |
FKS10080601 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base | PK |
NNC347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base | Pack |
377267 also known as NNC377267 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base | BAG |
FIS12-565-167N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base | PK |
FKS11341675 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base | PK |
368632 also known as NNC368632 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface | PK |
FIS12-565-171N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface | PK |
FKS10674511 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface | PK |
NNC368632 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface | Pack |
363401 | Round | 1.8 mL | Internal | Writing Surface | PK |
FKS10004220 | Round | 1.8 mL | Internal | Writing Surface | PK |
351934 also known as 35193 4 | Round | 1.8 mL | Internal | Writing Surface, No Cap | PK |
379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base | CS |
FIS12-565-162N | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base | PK |
NNC379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base | Pack |
FKS10789871 | Round | 3.6 mL | Internal | Writing Surface | PK |
337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base | PK |
FIS12-565-291 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base | PK |
NNC337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base | Pack |
FIS12-565-161N | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base | PK |
FKS10716922 | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base | PK |
363452 | Round | 4.5 mL | Internal | Writing Surface | CS |
FIS12-565-173N | Round | 4.5 mL | Internal | Writing Surface | PK |
FKS10358131 | Round | 4.5 mL | Internal | Writing Surface | PK |