Nunc™ TripleFlask Treated Cell Culture Flasks
Nunc™ TripleFlask Treated Cell Culture Flasks
Actual product may vary
Nunc™ TripleFlask Treated Cell Culture Flasks
Perform large cell mass applications such as drug screening and scale up with these cell-culture treated TripleFlasks.
Have Questions?
Catalog NumberSurface TreatmentClosure TypeBarcode
NNC132913Cell Culture-treatedFilterNo
Catalog number NNC132913
Price (SGD)
-
Surface Treatment:
Cell Culture-treated
Closure Type:
Filter
Barcode:
No
Perform cell culture expansion and experience 500 cm2 total growth area within the footprint of a standard flask using Thermo Scientific Nunc TripleFlask treated cell culture flasks. Total culture area is achieved via three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask cell culture flasks receive the Nunclon Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
Three layers of growth surface provide a total culture area of 500 cm2 in the same footprint as a standard 175 cm2 flask.
  • Recommended working volume is 200 mL
  • Promotes conventional, flat, monolayer culturing
  • External dimensions of 175 cm2
  • Also offered barcoded with large Code 128
Includes Nunclon Delta Certification with testing on four different cell lines.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
BarcodeNo
DescriptionCell Culture Treated TripleFlasks
Capacity (Metric)500 cm²
MaterialPolystyrene
Neck StyleStraight
No. per Case32/Cs.
No. per Pack1 Ea.
Product TypeFlask
ShapeSame external dimensions as the standard Nunc T175 with straight neck
DisposableSingle-use
GraduatedYes
SterilitySterile
Bottom ShapeFlat
Closure TypeFilter
Culture Area500 cm²
Surface TreatmentCell Culture-treated
Volume (Metric) Working200 mL
Unit SizeCase