Fixogum Rubber Cement
Actual product may vary
MP Biomedicals
Fixogum Rubber Cement
Made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.
Have Questions?
Catalog NumberQuantity
MPB11FIXO0125125 mL
Catalog number MPB11FIXO0125
Price (SGD)
144.00
Each
Quantity:
125 mL
Price (SGD)
144.00
Each
Applications: Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridization for FISH.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
FormatTube
Product TypeBiochemicals
Quantity125 mL
FormLiquid
Unit SizeEach
Contents & Storage
Room Temperature