Check Order Status
Taq Pol Bulk
MP Biomedicals
Taq Pol Bulk
For life science applications
Have Questions?
Change view
Catalog Number
Quantity
MPB11EPTQA100
1000U
MPB11EPTQA025
250U
2 Options
Catalog number
MPB11EPTQA100
Price (SGD)
1,321.00
Each
Add to cart
Quantity:
1000U
Price (SGD)
1,321.00
Each
Add to cart
Ready for CE IVD labeling of clinical applications
For Resaerch Use Only
Specifications
Concentration
5 U/μL
Description
Taq Pol Bulk
Quantity
1000U
Includes
Taq
DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl
2