Lysing Matrix B, 2 mL Tubes
Lysing Matrix B, 2 mL Tubes
Actual product may vary
Lysing Matrix B, 2 mL Tubes
Lysing Matrix B, 2 mL Tubes
Lysing Matrix B, 2 mL Tubes
MP Biomedicals
Lysing Matrix B, 2 mL Tubes
Lysing Matrix B contains 0.1 mm silica spheres for lysing bacteria and spores.
Have Questions?
Catalog NumberQuantity
MPB11691105050 x 2 mL Tubes
Catalog number MPB116911050
Price (SGD)
174.00
PK
Add to cart
Quantity:
50 x 2 mL Tubes
Price (SGD)
174.00
PK
Add to cart
Lysing Matrix B tubes contain 0.1 mm silica spheres and are used with FastPrep instruments or similar bead beaters to lyse bacteria, fungal tissues and spores.
  • Tube Size: 2 mL
  • Sample Types: Gram positive and negative bacteria, fungal tissues, spores
  • Bead material: 0.1 mm silica spheres
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Bead TypeSilica Spheres
Sample TypeBacteria, Fungi, Soil, Animal Tissues
DNaseDnase-free
For Use With (Equipment)FastPrep instruments or other bead beaters
For Use With (Application)Nucleic Acid Isolation and Purification, Homogenization
MatrixLysing Matrix B
Quantity50 x 2 mL Tubes
RNaseRNase-free
Unit SizePK
Contents & Storage
Store at Room Temperature.