Product Image
MP Biomedicals
SD MEDIUM-URA, Powder
Complete synthetic defined single dropout medium for the growth of S. cereviseae
Have Questions?
Catalog number MPB114813065
Price (SGD)
223.00
Each
Price (SGD)
223.00
Each
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Certifications/ComplianceMolecular biology Certified™
FormPowder
Product TypeYeast Media
FormulationYNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L, CSM-Ura 0.69g/L
For Use With (Application)For growth of Saccharomyces cerevisiae
Packaging TypePouch
Quantity10 x 500 mL
Shelf Life4 Years
Unit SizeEach
Contents & Storage
15°C to 30°C