YPD (YEPD) Broth ( Powder)
YPD (YEPD) Broth ( Powder)
Actual product may vary
MP Biomedicals
YPD (YEPD) Broth ( Powder)
Rich medium for the growth of S. cerevisiae in liquid medium
Have Questions?
Catalog NumberQuantity
MPB1140010321000 g
Catalog number MPB114001032
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
  • Contains, dextrose, peptone, and yeast extract
  • pH 6.5 at 300°C
  • Bulk powder packaging
Formulation
  • 50g/L
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose
Yeast Cell Culture
Specifications
Certifications/ComplianceMolecular biology Certified™
FormPowder
GradeMolecular biology
Product TypeMicrobiology Media
FormulationCasein Peptone type-M 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application)For growth of Saccharomyces cerevisiae in liquid medium
Packaging TypeBulk Powder
Quantity1000 g
Shelf Life4 Years
Unit SizeEA
Contents & Storage
15°C to 30°C