Casamino Acids
Actual product may vary
MP Biomedicals
Casamino Acids
Supplemented to microbiological growth media.
Have Questions?
Catalog NumberQuantity
MPB113060022454 g
Catalog number MPB113060022
Price (SGD)
937.00
Each
Quantity:
454 g
Price (SGD)
937.00
Each
Applications: Casamino acids is supplemented to microbiological growth media.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
Specifications
FormPowder
Product TypeMedia Supplement
Quantity454 g
Unit SizeEach
Contents & Storage
Store at Room Temperature (15-30 °C).