▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Additional Cell Culture Reagents and Supplements
▸
Glycerol, 80% Sterile Solution
Actual product may vary
MP Biomedicals
Glycerol, 80% Sterile Solution
Excellent for freezing bacterial and yeast strains
Have Questions?
Catalog Number
Quantity
MPB113055034
100 mL
Catalog number
MPB113055034
Price (SGD)
81.00
Each
Add to cart
Quantity:
100 mL
Price (SGD)
81.00
Each
Add to cart
Specifications
Product Type
Glycerol
Sterility
Sterile
Quantity
100 mL
Unit Size
Each
Contents & Storage
15° to 30°C