Check Order Status
LB-Agar, Lennox, Large Capsules
Actual product may vary
MP Biomedicals
LB-Agar, Lennox, Large Capsules
Culture media used for the growth of bacteria.
Have Questions?
Catalog number
MPB113002326
Price (SGD)
468.00
EA
Add to cart
Price (SGD)
468.00
EA
Add to cart
Typically contain 10g of tryptone and 5g of yeast extract per liter, and vary in salt
Specifications
Broth Type
LB Broth
Format
Large capsules
Media Type
Bacterial media
Product Type
Agar
Quantity
454 g