LB Medium, Dehydrated: Bulk Powder
Actual product may vary
MP Biomedicals
LB Medium, Dehydrated: Bulk Powder
Formulations of the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; contains 10g NaCl per liter
Have Questions?
Catalog NumberQuantity
MPB1130020321000 g
Catalog number MPB113002032
Price (SGD)
391.00
Each
Quantity:
1000 g
Price (SGD)
391.00
Each
Specifications
Certifications/ComplianceMolecular Biology Certified™
FormDehydrated
GradeMolecular Biology
Media TypeLB Medium
Product TypeDehydrated Culture Media
FormulationTryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
For Use With (Application)For growth of bacteria
Packaging TypeBulk Powder
Quantity1000 g
Unit SizeEach
Contents & Storage
15°C to 30°C