Geneclean™ Turbo Kit
Actual product may vary
MP Biomedicals
Geneclean™ Turbo Kit
Used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. MP Biomedicals™ Geneclean™ Turbo Kit can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.
Have Questions?
Catalog NumberNo. of Reactions
MPB111102600300 Preps
Catalog number MPB111102600
Price (SGD)
1,027.00
Each
No. of Reactions:
300 Preps
Price (SGD)
1,027.00
Each
  • Process with either vacuum or centrifugation and save time with patented Turbo filter design
  • Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
  • Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications
Research Use Only (RUO)
Specifications
FormatGeneclean Imbedded Turbo Spin Column
For Use With (Application)Nucleic Acid isolation and purification
No. of Reactions300 Preps
Unit SizeEach
Contents & Storage
Room Temperature