7X™-PF-O-Matic Detergent
7X™-PF-O-Matic Detergent
Actual product may vary
MP Biomedicals
7X™-PF-O-Matic Detergent
7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Have Questions?
Catalog NumberVolume (English)
MPB0976675944 x 1 gal.
Catalog number MPB097667594
Price (SGD)
425.00
EA
Add to cart
Volume (English):
4 x 1 gal.
Price (SGD)
425.00
EA
Add to cart
  • Environmentally safe and phosphate free
  • Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available
  • Stable for long-term storage
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
Specifications
FormLiquid
pH7 to 8
AppearanceDark Yellow
Concentration7X
For Use With (Application)Use on Glass, Plastic, automatic washers
Quantity4 Pk.
Volume (English)4 x 1 gal.
Unit SizeEA
Contents & Storage
15°C to 30°C