ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Have Questions?
Change view
Catalog Number
Volume (English)
MPB097667194
4 x 1 gal.
MPB097667193
1 gal.
2 Options
Catalog number MPB097667194
Price (SGD)
461.00
Each
Volume (English):
4 x 1 gal.
Price (SGD)
461.00
Each
Environmentally safe and phosphate free.
Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
Several convenient package sizes are available
Sold in concentrated form so the customer can prepare the appropriate dilution as needed
Completely drains from glassware in seconds
Eliminates residue leaving no interfering fluorescence
pH neutral, and no need for pH adjustment at any concentration
No etch to glass or plastic labware at any concentration
Specifications
FormLiquid
pH7 to 8
AppearanceYellow
Concentration7X
For Use With (Application)Use on Glass, Plastic, benchtop equipment and tubing