7X™ Laboratory Detergents
7X™ Laboratory Detergents
Actual product may vary
MP Biomedicals
7X™ Laboratory Detergents
Safe, effective cleaning solutions for glass and plastic labware
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)
MPB0976670931 gal.
MPB0976670944 x 1 gal.
Catalog number MPB097667093
Price (SGD)
93.00
Each
Add to cart
Volume (English):
1 gal.
Price (SGD)
93.00
Each
Add to cart
7X cleaning solutions are ideal for glass and plastic labware. 7X products contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware. The environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds.
7X Laboratory Detergents
  • For manual washing, phosphate-free
  • Comprised of concentrated liquids with an anionic-surface active agent and special solvents—compatible with alkalis and acids
  • A 5% solution in distilled water has a 6 to 9pH range with alkaline salts added as builders
7X-O-Matic™ Automatic Washer Detergents
  • For automatic or mechanical washers—effective in temperatures from ambient to 85°C (185°F)
  • Highly concentrated, stabilized, and buffered solution with a unique non-ionic surfactant that is stable to acids and bases, phosphate-free
  • Contain a powerful wetting agent to clean microscopic crevices not normally accessible to ordinary detergents
  • A 1% solution in distilled water has a 9 to 11pH range
Specifications
FormLiquid
pH6 to 7.5
Concentration7X
For Use With (Application)Use on Glass, Plastic, benchtop equipment and tubing
Volume (English)1 gal.
Unit SizeEach
Contents & Storage
15°C to 30°C