Serum, Type AB, Human, From Male Donors
Serum, Type AB, Human, From Male Donors
Actual product may vary
MP Biomedicals
Serum, Type AB, Human, From Male Donors
Used as a growth supplement for cell culture media and in other research applications.
Have Questions?
Catalog NumberQuantity
MPB092930949100 mL
Catalog number MPB092930949
Price (SGD)
520.00
Each
Add to cart
Quantity:
100 mL
Price (SGD)
520.00
Each
Add to cart
  • Collected from type AB human blood
  • Used as a growth supplement for cell culture media and in other research applications
  • Proven to grow many human cell lines at a faster rate than mixed blood group serum
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Certifications/ComplianceU.S. regulations
ColorStraw Yellow to Golden Brown
SpeciesHuman
FormLiquid
Product TypeHuman Serum
Quantity100 mL
SterilitySterile
Sterilization Method0.1 μm filtration
For Use With (Application)Grow many Human cell lines at a faster rate
Hemoglobin Concentration≤26 mg/dL
Mycoplasma DetectedNo
Osmolality263 to 323 mOsm/kg
pH7 to 8
Protein Content5 to 8.3 g/dL
Unit SizeEach
Contents & Storage
-20°C