Chicken Embryo Extract
Actual product may vary
MP Biomedicals
Chicken Embryo Extract
Chicken Embryo Extract, cell culture supplement
Have Questions?
Catalog NumberQuantity
MPB09285014520 mL
Catalog number MPB092850145
Price (SGD)
758.00
Each
Quantity:
20 mL
Price (SGD)
758.00
Each
Chicken Embryo Extract is obtained from ten-day-old embryos. The product is intended for in vitro research use only.
  • Chicken embryo extract is prepared from 9 to 12 day old eggs collected from registered flocks and found to be free from clinical signs of notifiable diseases
  • Widely used as cell culture supplement for avian neural crest cells, muscle cells, dorsal root ganglion neurons, etc
  • Supplemented with 100 IU/mL Penicillin, 100 μg/mL Streptomycin and 2.5 μg/mL Amphotericin, and sterile filtered to 0.2 μm
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
DescriptionChick Embryo Extract
FormFrozen
Product TypeGrowth Factor
SterilitySterile
Sterilization Method0.2 μm Sterile-filtered
FormulationSupplied in Dulbecco's PBS
For Use With (Application)Growth factor supplement in tissue culture
Quantity20 mL
Regulatory StatusRUO
Unit SizeEach
Contents & Storage
Store at -20°C.