Check Order Status
Murashige and Skoog Media
Actual product may vary
MP Biomedicals
Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Have Questions?
Catalog Number
Quantity
Product Type
Content And Storage
MPB092623122
1 x 10 L
Murashige and Skoog Basal Medium
Catalog number
MPB092623122
Price (SGD)
68.00
Each
Add to cart
Quantity:
1 x 10 L
Product Type:
Murashige and Skoog Basal Medium
Price (SGD)
68.00
Each
Add to cart
Each formulation is prepared according to the original specification, except where noted
Finished lots of media are plant cell tested
Specifications
Form
Powder
Product Type
Murashige and Skoog Basal Medium
Quantity
1 x 10 L
Format
Solution
pH
3.75 to 4.75
Unit Size
Each
Contents & Storage
4°C