LymphoSep™ Lymphocyte Separation Medium
Actual product may vary
MP Biomedicals
LymphoSep™ Lymphocyte Separation Medium
LymphoSep™ Lymphocyte Separation Medium is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinized or defibrinated human blood.
Have Questions?
Catalog NumberQuantity
MPB091692249100 mL
Catalog number MPB091692249
Price (SGD)
81.00
Each
Quantity:
100 mL
Price (SGD)
81.00
Each
  • Lymphocytes are isolated by layering diluted, heparinized blood over a solution of sodium metrizoate that contains a polysaccharide
  • Polysaccharide aggregates the erythrocytes so that they can be sedimented by low centrifugation
  • The less dense lymphocytes remain as a clearly defined band at the plasma Lymphosep interface
  • Provides:
    • In vitro isolation of lymphocytes from diluted whole blood
    • Maximum yield of mononuclear cells
    • Easy one-step centrifugation
    • >96% cell viability of lymphocytes
    • Low endotoxins
    • Autoclave sterilization
Specifications
Product TypeLymphosep Lymphocyte Separation Medium
SterilitySterile
pH6 to 9
Quantity100 mL
Unit SizeEach
Contents & Storage
15° to 30°C