Min. Ess. Med. Eagle Non-Ess. Amino Acids
Min. Ess. Med. Eagle Non-Ess. Amino Acids
Actual product may vary
MP Biomedicals
Min. Ess. Med. Eagle Non-Ess. Amino Acids
For use in cell culture applications using defined media for the in vitro cultivation of cells
Have Questions?
Catalog NumberQuantity
MPB091681049100 mL
Catalog number MPB091681049
Price (SGD)
57.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
57.00
EA
Add to cart
Specifications
Product TypeAmino Acid Supplement
SterilitySterile
Without AdditivesNo Glutamine
Quantity100 mL
pH2.8 to 3.8
Unit SizeEA
Contents & Storage
4°C