▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Antibiotics
▸
G418 Sulfate Solution, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
G418 Sulfate Solution, MP Biomedicals
50mg/ml active antibiotic sterile-aqueous solution. Purity: >>98%
Have Questions?
Catalog Number
Quantity
MPB091672546
20 mL
Catalog number
MPB091672546
Price (SGD)
347.00
Each
Add to cart
Quantity:
20 mL
Request bulk or custom format
Price (SGD)
347.00
Each
Add to cart
Specifications
Chemical Name or Material
G418 Sulfate Solution
Type
G418 Sulfate
Quantity
20 mL