▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Media
▸
Classical and Basal Cell Culture Media
▸
RPMI Cell Culture Media
▸
RPMI 1640 Medium
Actual product may vary
MP Biomedicals
RPMI 1640 Medium
General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes
Have Questions?
Catalog Number
pH
Content And Storage
MPB091646754
6.8 to 7.4
Catalog number
MPB091646754
Price (SGD)
67.00
Each
Add to cart
pH:
6.8 to 7.4
Price (SGD)
67.00
Each
Add to cart
Specifications
Product Type
RPMI 1640 Medium
Sterility
Sterile
Without Additives
No Glutamine, No Phenol Red
Quantity
500 mL
pH
6.8 to 7.4
Unit Size
Each
Contents & Storage
0° to 5°C