Tryptic Soy Agar
Actual product may vary
MP Biomedicals
Tryptic Soy Agar
Used for the cultivation of fastidious and non-fastidious microorganisms.
Have Questions?
Catalog NumberQuantity
MPB091010617500 g
Catalog number MPB091010617
Price (SGD)
206.00
Each
Quantity:
500 g
Price (SGD)
206.00
Each
Tryptic Soy Agar is a medium very rich in nutrients for 'general use' in microbiological laboratories.
Containing two peptones obtained by enzymatic hydrolysis of casein and soy protein, this medium supports the growth of a great variety of microorganisms, including fastidious aerobes and anaerobes.
If desired, antibiotics can easily be incorporated as well as other supplements or inhibitory agents.
A short list of microorganisms that grow on this medium are the following: Streptococcus, Neisseria, Brucella, Corynebacterium, Listeria, Pasteurella, Vibrio, Haemophilus vaginals, Candida, etc.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
FormPowder
Media TypeTryptic Soy Agar
Product TypeTryptic Soy Agar
FormulationCasein Peptone 15 g/L, Sodium Chloride 5 g/L, Soy Peptone 5 g/L, Agar 15 g/L
For Use With (Application)Isolation and cultivation of fastidious organisms
Quantity500 g
Unit SizeEach
Contents & Storage
15 to 30°C