γ-Globulin, Human Plasma, 1g
Actual product may vary
MP Biomedicals
γ-Globulin, Human Plasma, 1g
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2, HBsAg, Anti-HCV and HIV-1 Ag by FDA approved methods or CE approved methods.
Have Questions?
Catalog NumberQuantity
MPB088231011 g
Catalog number MPB08823101
Price (SGD)
271.00
Each
Quantity:
1 g
Price (SGD)
271.00
Each
Specifications
ColorWhite
pH6.44
Quantity1 g
FormPowder
For Use With (Application)Used as an immunological challenge in studies of mechanisms of immunotolerance
Purity or Quality Grade>96%
Product TypeHuman Plasma gamma Globulin
Unit SizeEach
Contents & Storage
2°C to 8°C