Albumin, Bovine, IVD Grade, 98%
Actual product may vary
MP Biomedicals
Albumin, Bovine, IVD Grade, 98%
Prepared by Cohn fractionation
Have Questions?
Catalog NumberQuantity
MPB08820451100g
Catalog number MPB08820451
Price (SGD)
1,243.00
Each
Quantity:
100g
Price (SGD)
1,243.00
Each
  • Protease-, peroxidase- and phosphatase-free
  • Purity: >98%
  • pH: 7
Specifications
For Use With (Application)Immunoassay, Mammalian Cell Culture
NameBovine Albumin Cohn Fraction V
pH Range7
Preparation MethodPlasma undergoes salt fractionation using Caprylic Acid (Sodium Salt)
Purification MethodSalt Fractionation
Quantity100g
Storage Requirements2° to 8°C