Check Order Status
Albumin, Bovine, Reagent Grade
Actual product may vary
MP Biomedicals
Albumin, Bovine, Reagent Grade
Prepared by heat shock fractionation
Have Questions?
Catalog Number
Quantity
MPB08810661
50g
Catalog number
MPB08810661
Price (SGD)
428.00
EA
Add to cart
Quantity:
50g
Price (SGD)
428.00
EA
Add to cart
For IVD assays, immunoassays, enzyme stabilization
Low fatty acids, IgG
Purity: >98%
pH: 7
Specifications
Concentration
1mg/mL
For Use With (Application)
Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol)
66
Name
Bovine Albumin Fraction V
pH Range
7
Quantity
50g
Storage Requirements
0° to 5°C