Check Order Status
Mounting Medium, Immuno-Fluore
Actual product may vary
MP Biomedicals
Mounting Medium, Immuno-Fluore
designed for immunofluorescent preparations
Have Questions?
Catalog Number
Quantity
MPB08622701
25 mL
Catalog number
MPB08622701
Price (SGD)
887.00
EA
Add to cart
Quantity:
25 mL
Price (SGD)
887.00
EA
Add to cart
Specifications
Product Type
Immuno-Fluore mounting medium
Quantity
25 mL
For Use With (Application)
Immunofluorescence (IF)
Form
Liquid
Unit Size
EA
Contents & Storage
15° to 30°C