Check Order Status
LSM™ Lymphocyte Separation Medium
Actual product may vary
MP Biomedicals
LSM™ Lymphocyte Separation Medium
LSM™ Lymphocyte Separation Medium is a high performance density gradient medium for monocyte separation.
Have Questions?
Catalog Number
Quantity
MPB0850494X
100 mL
Catalog number
MPB0850494X
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
LSM is a sterile-filtered solution of a sucrose polymer and diatrizoate salts
Formulated for isolation of mononuclear cells from defibrinated or heparinized whole human blood
One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
Maximum yield of mononuclear cells
> 96% cell viability of lymphocytes
Easy one-step centrifugation
Low endotoxins
Autoclave sterilization
For Research Use Only.
Specifications
Form
Liquid
Product Type
Lymphocyte Separation Medium
Sterility
Sterile
For Use With (Application)
Cell isolation, expansion
Quantity
100 mL
pH
7.23
Unit Size
EA
Contents & Storage
Store at Room temperature(15° to 30°C)