Zymolyase 20T Powder, 1 g
Zymolyase 20T Powder, 1 g
Actual product may vary
MP Biomedicals
Zymolyase 20T Powder, 1 g
Zymolyase 20T from Anthrobacter luteus in lyophilized powder form.
Have Questions?
Catalog number MPB08320921
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Zymolyase 20T from Anthrobacter luteus in lyophilized powder form.
Zymolyase is used for lysing the cell walls of viable yeast cells and preparing yeast spheroblasts. This zymolyase is a highly-purified preparation from Arthrobacter luteus. One unit is defined as the amount of activity causing a decrease in A800 of 30%.
  • Lyophilized powder
  • Should be stored at 2°C to 8°C
  • Efficiently hydrolyzes poly (β-1,3 glucose)-like yeast cell wall glucan
  • Lytic activity of 20,000 U/g
Specifications
Donor SourceArthrobacter luteus
FormLyophilized
For Use With (Application)Lysing Yeast Cell Walls to Produce Protoplast or Spheroplast
EnzymeZymolyase
Quantity1 g
Unit SizeEA