DNA-Erase™ Decontaminant
DNA-Erase™ Decontaminant
Actual product may vary
MP Biomedicals
DNA-Erase™ Decontaminant
Removes DNA contamination
Have Questions?
Catalog NumberVolume (Metric)
MPB04821805500 mL
Catalog number MPB04821805
Price (SGD)
136.00
EA
Add to cart
Volume (Metric):
500 mL
Price (SGD)
136.00
EA
Add to cart
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination. Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. By following the simple decontamination instructions, DNA is completely destroyed and removed.
  • Eliminates PCR artifacts from all laboratory surfaces and apparatus, including pipetters
  • Stable, heat-resistant, and noncorrosive
  • Supplied as ready-to-use formula
Specifications
Product TypeDecontaminant Solution
Volume (English)16.9 oz.
Volume (Metric)500 mL
For Use With (Application)Removal of DNA contamination
Unit SizeEA