Glycine, Electrophoresis Grade, MP Biomedicals™
Glycine, Electrophoresis Grade, MP Biomedicals™
Glycine, Electrophoresis Grade, MP Biomedicals™
Glycine, Electrophoresis Grade, MP Biomedicals™
MP Biomedicals
Glycine, Electrophoresis Grade, MP Biomedicals™
Glycine, Electrophoresis Grade is for use in electrophoresis buffers and SDS-PAGE.
Have Questions?
Catalog NumberQuantity
MPB048088221 kg
Catalog number MPB04808822
Price (SGD)
175.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
175.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycine
Melting Point451°F
CAS56-40-6
pH4 (0.2 molar aq. soln.)
Molecular FormulaC2H5NO2
View more
  • Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
  • Glycine is also a component of Towbin's transfer buffer for Western blots.
  • It is an inhibitory neurotransmitter in the spinal cord and an allosteric regulator of NMDA receptors.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.