Check Order Status
Tricine, Electrophoresis Grade, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Tricine, Electrophoresis Grade, MP Biomedicals
5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer
Have Questions?
Catalog Number
Quantity
MPB04807420
1 kg
Catalog number
MPB04807420
Price (SGD)
1,123.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
1,123.00
EA
Add to cart
Specifications
Chemical Name or Material
Tricine
CAS
4-1-5704
Quantity
1 kg
Molecular Formula
C6H13NO5
InChI Key
SEQKRHFRPICQDD-UHFFFAOYSA-N
View more