Ethidium Bromide Solution, MP Biomedicals
Ethidium Bromide Solution, MP Biomedicals
Ethidium Bromide Solution, MP Biomedicals
Ethidium Bromide Solution, MP Biomedicals
MP Biomedicals
Ethidium Bromide Solution, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB0480251110 mL
Catalog number MPB04802511
Price (SGD)
113.00
EA
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (SGD)
113.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthidium Bromide Solution
CAS1239-45-8
Molecular FormulaC21H20BrN3
MDL NumberMFCD00011724
Synonymethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
View more