Check Order Status
Glycerol (Ultra Pure 99.5%)
Element Search
Structure Search
MP Biomedicals
Glycerol (Ultra Pure 99.5%)
Have Questions?
Catalog Number
Quantity
MPB04800687
500 mL
Catalog number
MPB04800687
Price (SGD)
147.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
147.00
EA
Add to cart
Specifications
Chemical Name or Material
Glycerol (Ultra Pure 99.5%)
CAS
56-81-5
Molecular Formula
C3H8O3
MDL Number
MFCD00004722
Quantity
500 mL
View more