Z-VAD(OMe)-Fluromethylketone Inhibitor
Z-VAD(OMe)-Fluromethylketone Inhibitor
Actual product may vary
MP Biomedicals
Z-VAD(OMe)-Fluromethylketone Inhibitor
The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.
Have Questions?
Catalog NumberQuantity
MPB03FK009011 mg
Catalog number MPB03FK00901
Price (SGD)
436.00
EA
Add to cart
Quantity:
1 mg
Price (SGD)
436.00
EA
Add to cart
Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).
Specifications
ColorWhite
Quantity1 mg
Product TypeZ-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor
Molecular Weight (g/mol)467.487
FormSolid
Unit SizeEA
Contents & Storage
4°C