Choline Deficient Diet, Pellet
Actual product may vary
MP Biomedicals
Choline Deficient Diet, Pellet
Used for the study characterisation of a stereotypical cellular and extracellular adult liver progenitor cell niche in rodents and diseased human liver.
Have Questions?
Catalog NumberQuantity
MPB029600341010 kg
Catalog number MPB0296003410
Price (SGD)
1,055.00
Each
Quantity:
10 kg
Price (SGD)
1,055.00
Each
Composition (Ingredient, Amount):
  • Vitamin Free Casei,n 10%
  • a-Protein, 10%
  • Sucrose, 56%
  • Lard, 20%
  • Salt Mixture Wesson, 4%
  • Calcium Carbonate, 21% (of total salt mixture)
  • Copper Sulfate.5H2O, 0.039% (of total salt mixture)
  • Ferric Phosphate, 1.47% (of total salt mixture)
  • Manganese Sulfate.H2O, 0.02% (of total salt mixture)
  • Magnesium Sulfate.7H2O, 9% (of total salt mixture)
  • Potassium Aluminum Sulfate, 0.009% (of total salt mixture)
  • Potassium Chloride, 12% (of total salt mixture)
  • Potassium Dihydrogen Phosphate, 31% (of total salt mixture)
  • Potassium Iodide, 0.005% (of total salt mixture)
  • Sodium Chloride, 10.5% (of total salt mixture)
  • Sodium Fluoride, 0.057% (of total salt mixture)
  • Tricalcium Phosphate, 14.9% (of total salt mixture)
  • Plus MP Vitamin Diet Fortification Mixture except Choline Chloride
Specifications
Product TypeAnimal Diet
For Use With (Application)Research animal diet nutrient
Quantity10 kg
Storage Requirements4°C
Unit SizeEach