▸
Biochemical Reagents
▸
Carbohydrates
▸
Glycosyl compounds
▸
Sucrose, (Crystalline), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sucrose, (Crystalline), MP Biomedicals
This is a slightly less pure sucrose than our 190271. It does not reduce Fehling's solution, and is suitable for most routine applications
Have Questions?
Catalog Number
Quantity
MPB0290297825
25 lb.
Catalog number
MPB0290297825
Price (SGD)
248.00
Each
Add to cart
Quantity:
25 lb.
Request bulk or custom format
Price (SGD)
248.00
Each
Add to cart
Specifications
Chemical Name or Material
Sucrose
CAS
57-50-1
Recommended Storage
Room Temperature
Molecular Formula
C12H22O11
MDL Number
MFCD00006626
View more
Its a simple carbohydrate.
It is used in density gradients, Dielectrophoretic Field-Flow Fractionation running buffer.
It is a main component of media and animal diet.
Sucrose is a simple carbohydrate, a disaccharide composed of glucose and fructose.
It is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme