▸
Organic compounds
▸
Unclassified Organic Compounds
▸
Doxycycline Hyclate USP MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Doxycycline Hyclate USP MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219895510
10 g
Catalog number
MPB0219895510
Price (SGD)
467.00
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (SGD)
467.00
Each
Add to cart
Specifications
Chemical Name or Material
Doxycycline Hyclate
CAS
24390-14-5
Molecular Formula
C26H41ClN2O12
Quantity
10 g
Synonym
doryx, doxycycline hyclate, doxycycline, hydrochloride, hemiethanolate, hemihydrate;
View more