1x1 image pixel for data collection
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
MP Biomedicals
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
100 mL
0.5 L
Catalog number MPB0219605591
Price (SGD)
311.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
311.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl sulfoxide
CAS67-68-5
Quantity1 L
Physical FormLiquid
Assay Percent Range≥99%
View more
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link