Vancomycin hydrochloride, MP Biomedicals™
Vancomycin hydrochloride, MP Biomedicals™
Vancomycin hydrochloride, MP Biomedicals™
MP Biomedicals
Vancomycin hydrochloride, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB02195540055 g
Catalog number MPB0219554005
Price (SGD)
1,190.00
Each
Quantity:
5 g
Request bulk or custom format
Price (SGD)
1,190.00
Each
Specifications
Chemical Name or MaterialVancomycin hydrochloride
TypeVancomycin Hydrochloride
Physical FormPowder
CAS123409-00-7