Check Order Status
Propidium Iodide, 99%, For HPLC analysis, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Propidium Iodide, 99%, For HPLC analysis, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219545825
25 mg
Catalog number
MPB0219545825
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Specifications
Chemical Name or Material
Propidium Iodide
Name Note
For HPLC analysis
CAS
25535-16-4
Color
Red
Physical Form
Flaky Powder
View more