Pepsin
Product Image
MP Biomedicals
Pepsin
From porcine stomach mucosa
Have Questions?
Catalog NumberQuantity
MPB021953671010 g
Catalog number MPB0219536710
Price (SGD)
1,768.00
EA
Add to cart
Quantity:
10 g
Price (SGD)
1,768.00
EA
Add to cart
Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.
Specifications
For Use With (Application)Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies
pH Range1 to 3.5
Sample TypePorcine Stomach Mucosa
FormLyophilized
Quantity10 g
Product TypePepsin
Unit SizeEA
Contents & Storage
0°C