β-Glucosidase
Actual product may vary
MP Biomedicals
β-Glucosidase
From Sweet Almonds
Have Questions?
Catalog NumberQuantity
MPB0219519780100 kU
Catalog number MPB0219519780
Price (SGD)
-
Quantity:
100 kU
Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.
Specifications
Concentration≥1000 U/mg
ColorWhite
For Use With (Application)Synthesis of glucosides and fucosides
SourceSweet almonds
Quantity100 kU
Product Typebeta-Glucosidase
FormPowder
Unit SizeEach
Contents & Storage
-20°C