1x1 image pixel for data collection
Carbenicillin Disodium Salt, MP Biomedicals™
Carbenicillin Disodium Salt, MP Biomedicals™
Carbenicillin Disodium Salt, MP Biomedicals™
Carbenicillin Disodium Salt, MP Biomedicals™
MP Biomedicals
Carbenicillin Disodium Salt, MP Biomedicals™
Displays antipseudomonal and antiproteal effects. Carbenicillin Disodium Salt, MP Biomedicals is a carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface
Have Questions?
Catalog NumberQuantity
MPB02195092.2250 mg
Catalog number MPB02195092.2
Price (SGD)
90.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (SGD)
90.00
EA
Add to cart
Specifications
Physical FormPowder
CAS4800-94-6
Recommended StorageStore at 4°C, desiccate.
Potency>650μg/mg
ConcentrationWorking Concentration: 105u/L (5-50μg/mL) in cell culture; approximately 200mg/kg in vivo.
View more
Carbenicillin is a semi-synthetic antibiotic that is an analog to ampicillin. It is derived from benzyl penicillin by substituting the a-carboxyl group in the benzyl side chain. It is completely ionized in the blood and is about 50% reversibly protein bound in plasma. Its tissue distribution is similar to other penicillins.
Susceptible bacteria include gram-positive and gram-negative bacteria abd pseudomonas.Though it has substantial in vitro activity against a variety of both gram-positive and gram-negative microorganisms, the most important aspect of its profile is in its antipseudomonal and antiproteal effect.
Carbenicillin Disodium Salt is used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link