▸
Acids
▸
Inorganic Acids
▸
Boric Acid
▸
Boric Acid MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Boric Acid MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219507401
1 kg
Catalog number
MPB0219507401
Price (SGD)
138.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
138.00
Each
Add to cart
Specifications
Chemical Name or Material
Boric Acid
CAS
10043-35-3
Density
1.5172 g/cm3 at 14 °C
Quantity
1 kg
Molecular Formula
BH3O3
View more