Check Order Status
Sodium Perchlorate Monohydrate, 85-90%, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Perchlorate Monohydrate, 85-90%, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219486290
500 g
Catalog number
MPB0219486290
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Perchlorate Monohydrate
CAS
7791-07-3
Recommended Storage
Room Temperature
Quantity
500 g
Assay Percent Range
85-90%
View more