Check Order Status
MOPS, free acid, 100.2%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
MOPS, free acid, 100.2%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219483790
500 g
Catalog number
MPB0219483790
Price (SGD)
709.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
709.00
EA
Add to cart
Specifications
Chemical Name or Material
MOPS
CAS
1132-61-2
Quantity
500 g
Molecular Formula
C7H15NO4S
MDL Number
MFCD00006183
View more